Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253877.1 | NP_001240806.1 | 5 | 989 | ||
NM_001253878.1 | NP_001240807.1 | 5 | 708 | |||
NM_001253879.1 | NP_001240808.1 | 4 | 631 | |||
NM_001253880.1 | NP_001240809.1 | 2 | 346 | |||
NM_013827.3 | NP_038855.2 | 4 | 627 | |||
GenBank mRNA | AB050541.1 | - | 4 | 562 | ||
AB221622.1 | - | 1 | 34 | |||
AK030752.1 | - | 4 | 577 | |||
AK031409.1 | - | 6 | 696 | |||
AK032798.1 | - | 5 | 950 | |||
AK032819.1 | - | 5 | 951 | |||
AK137539.1 | - | 6 | 936 | |||
AK139685.1 | - | 5 | 709 | |||
BC024889.1 | - | 4 | 599 | |||
BC029076.1 | - | 4 | 538 | |||
BC050826.1 | - | 4 | 581 | |||
BC057163.1 | - | 4 | 424 | |||
BC071223.1 | - | 2 | 99 | |||
BC092237.1 | - | 4 | 559 | 30637248 | ||
BC100340.1 | - | 2 | 277 | 30715034 | ||
S78454.1 | - | 1 | 34 |