Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161775.1 | NP_001155247.1 | 37 | 5412 | ||
NM_013607.2 | NP_038635.2 | 37 | 5412 | |||
GenBank mRNA | AK173045.1 | - | 37 | 5376 | ||
D85923.1 | - | 37 | 5367 | |||
D85924.1 | - | 37 | 5367 |