Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177369.1 | NP_001170840.1 | 18 | 2209 | ||
NM_001177370.1 | NP_001170841.1 | 17 | 2125 | |||
NM_019408.3 | NP_062281.1 | 17 | 2360 | |||
GenBank mRNA | AF155372.1 | - | 18 | 2287 | ||
AF155373.1 | - | 18 | 2324 | |||
AK134081.1 | - | 18 | 2628 | |||
AK137680.1 | - | 18 | 2199 | |||
AK144994.1 | - | 18 | 2202 | |||
AK145312.1 | - | 17 | 2326 | |||
AK148160.1 | - | 18 | 2197 | |||
AK150012.1 | - | 18 | 2188 | |||
AK153412.1 | - | 13 | 1769 | |||
AK178267.1 | - | 1 | 62 | |||
BC027423.1 | - | 17 | 2126 | 3672913 | ||
DQ020182.1 | - | 17 | 2057 | |||
DQ020183.1 | - | 16 | 2178 | |||
DQ020184.1 | - | 15 | 2258 | |||
DQ020185.1 | - | 17 | 2057 |