Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008710.3 | NP_032736.2 | 458 | |||
GenBank mRNA | AK033282.1 | - | 3 | 399 | ||
AK040460.1 | - | 3 | 419 | |||
AK050478.1 | - | 2 | 363 | |||
AK052061.1 | - | 3 | 397 | |||
AK052234.1 | - | 3 | 443 | |||
AK054332.1 | - | 3 | 444 | |||
AK083413.1 | - | 3 | 444 | |||
AK084546.1 | - | 3 | 441 | |||
AK084604.1 | - | 3 | 441 | |||
AK085260.1 | - | 1 | 1852 | |||
AK085903.1 | - | 3 | 444 | |||
AK087064.1 | - | 3 | 440 | |||
AK088044.1 | - | 3 | 440 | |||
AK090115.1 | - | 461 | ||||
AK129006.1 | - | 3 | 435 | |||
AK159694.1 | - | 3 | 444 | |||
AK168736.1 | - | 2 | 241 | |||
BC008518.1 | - | 3 | 421 | 3601702 | ||
Z49204.1 | - | 2 | 365 |