Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Notch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010929.2 | NP_035059.2 | 30 | 6018 | ||
GenBank mRNA | AK143484.1 | - | 13 | 3184 | ||
BC138044.1 | - | 30 | 5976 | 8861063 | ||
BC145114.1 | - | 31 | 6106 | |||
M80456.1 | - | 30 | 5983 |