Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010990.1 | NP_035120.1 | 1 | 213 | ||
GenBank mRNA | AF102522.1 | - | 1 | 39 |