Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011019.3 | NP_035149.2 | 18 | 2842 | ||
GenBank mRNA | AB015978.1 | - | 18 | 3493 | ||
AF058805.1 | - | 18 | 2812 | |||
AK157508.1 | - | 13 | 2015 | |||
AK162684.1 | - | 1 | 144 | |||
BC137703.1 | - | 17 | 2740 | 8860722 |