Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011019.3 | NP_035149.2 | 7, 8 | 1103 | ||
GenBank mRNA | AB015978.1 | - | 7, 8 | 1757 | ||
AF058805.1 | - | 7, 8 | 1073 | |||
AK157508.1 | - | 2, 3 | 273 | |||
BC137703.1 | - | 6, 7 | 998 | 8860722 |