Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162946.1 | NP_001156418.1 | 18 | 2910 | ||
NM_008797.3 | NP_032823.2 | 17 | 2887 | |||
GenBank mRNA | AK077665.1 | - | 9 | 1715 | ||
AK128927.1 | - | 2684 | ||||
AK148328.1 | - | 18 | 2857 | |||
AK170596.1 | - | 18 | 2870 | |||
AK170871.1 | - | 18 | 2861 | |||
AK172320.1 | - | 18 | 2838 | |||
BC055030.1 | - | 18 | 2845 | 5053475 | ||
L09192.1 | - | 17 | 2863 |