Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163487.1 | NP_001156959.1 | 14 | 1418 | ||
NM_001163488.1 | NP_001156960.1 | 14 | 1354 | |||
NM_021514.4 | NP_067489.3 | 13 | 1371 | |||
GenBank mRNA | AF249894.1 | - | 13 | 1311 | ||
AK002711.2 | - | 14 | 1321 | |||
AK049773.1 | - | 14 | 1326 | |||
BC005526.1 | - | 13 | 1335 | 3495173 |