Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008866.2 | NP_032892.1 | 3, 4 | 199 | ||
GenBank mRNA | AK002674.1 | - | 3, 4 | 203 | ||
AK050549.1 | - | 3, 4 | 176 | |||
AK146874.1 | - | 3, 4 | 214 | |||
AK167231.1 | - | 221 | ||||
BC013536.1 | - | 3, 4 | 199 | 4240573 | ||
BC052848.1 | - | 3, 4 | 240 | 30055025 | ||
CT010201.1 | - | 3, 4 | 178 | |||
U89352.1 | - | 3, 4 | 178 |