Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008867.2 | NP_032893.1 | 22 | 3283 | ||
GenBank mRNA | AK036808.1 | - | 3 | 256 | ||
BC141355.1 | - | 22 | 3164 | 9055967 | ||
BC141356.1 | - | 22 | 3164 | 9055968 | ||
D30779.1 | - | 22 | 3236 |