Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011120.2 | NP_035250.1 | 4, 5 | 411 | ||
GenBank mRNA | AK005421.1 | - | 4, 5 | 408 | ||
AK005557.1 | - | 4, 5 | 411 | |||
AK019461.1 | - | 4, 5 | 411 | |||
BC099470.1 | - | 4, 5 | 400 | 30787403 | ||
X02594.1 | - | 4, 5 | 411 |