Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011163.4 | NP_035293.1 | 14 | 1510 | ||
GenBank mRNA | AK028602.1 | - | 14 | 1510 | ||
BC016422.1 | - | 14 | 1473 | 3964935 | ||
M65029.1 | - | 14 | 1462 | |||
M93567.1 | - | 14 | 1452 | |||
S48937.1 | - | 4 | 490 | |||
X62550.1 | - | 4 | 490 |