Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011204.2 | NP_035334.2 | 18 | 3022 | ||
GenBank mRNA | AK014577.1 | - | 17 | 2490 | ||
AK046854.1 | - | 4 | 624 | |||
AK080313.1 | - | 18 | 3022 | |||
BC057205.1 | - | 18 | 2990 | |||
D83966.1 | - | 18 | 2710 | |||
Z32740.1 | - | 17 | 2679 |