Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011211.3 | NP_035341.2 | 34 | 4599 | ||
GenBank mRNA | AF326559.1 | - | 16 | 3152 | ||
AF326560.1 | - | 16 | 3152 | |||
BC025145.1 | - | 10 | 1444 | |||
BC086654.1 | - | 25 | 3832 | |||
BC141409.1 | - | 24 | 3778 | 9056022 | ||
D13903.1 | - | 16 | 3549 |