Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpru | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083119.2 | NP_001076588.1 | 2, 3 | 332 | ||
NM_011214.2 | NP_035344.2 | 2, 3 | 332 | |||
GenBank mRNA | AK052720.1 | - | 2, 3 | 307 | ||
AK148515.1 | - | 2, 3 | 329 | |||
BC048694.1 | - | 2, 3 | 288 | |||
BC080736.1 | - | 2, 3 | 287 | |||
D88187.1 | - | 2, 3 | 323 |