Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163301.2 | NP_001156773.1 | 5 | 1073 | ||
NM_001163302.2 | NP_001156774.1 | 5 | 1069 | |||
NM_001163305.2 | NP_001156777.1 | 5 | 1152 | |||
NM_001163306.2 | NP_001156778.1 | 5 | 1071 | |||
NM_001267714.1 | NP_001254643.1 | 4 | 1005 | |||
NM_001267715.1 | NP_001254644.1 | 4 | 1009 | |||
NM_008994.4 | NP_033020.2 | 6 | 1216 | |||
GenBank mRNA | AF031128.1 | - | 5 | 1064 | ||
AK146530.1 | - | 5 | 981 | |||
AK151121.1 | - | 4 | 903 | |||
AK152331.1 | - | 4 | 903 | |||
BC012404.1 | - | 6 | 1096 | 3964491 |