Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 4 | 690 | ||
GenBank mRNA | AB232606.1 | - | 4 | 466 | ||
AK146452.1 | - | 4 | 551 | |||
AK146460.1 | - | 4 | 502 | |||
AK146623.1 | - | 4 | 502 | |||
AK146978.1 | - | 4 | 482 | |||
AK147077.1 | - | 4 | 521 | |||
AK148022.1 | - | 4 | 563 | |||
AK148106.1 | - | 4 | 489 | |||
AK188721.1 | - | 2 | 139 | |||
AK203890.1 | - | 2 | 164 | |||
BC054753.1 | - | 4 | 506 | 6411651 | ||
BC085270.1 | - | 4 | 499 | 6478784 | ||
BC157916.1 | - | 4 | 519 | 9007344 | ||
KJ650201.1 | - | 4 | 466 | |||
L26528.1 | - | 4 | 501 |