Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 1 | 245 | ||
GenBank mRNA | AB232606.1 | - | 1 | 21 | ||
AK134125.1 | - | 1 | 65 | |||
AK146452.1 | - | 1 | 106 | |||
AK146460.1 | - | 1 | 57 | |||
AK146623.1 | - | 1 | 57 | |||
AK146978.1 | - | 1 | 37 | |||
AK147077.1 | - | 1 | 76 | |||
AK148022.1 | - | 1 | 118 | |||
AK148106.1 | - | 1 | 44 | |||
AK186595.1 | - | 1 | 12 | |||
AK189626.1 | - | 1 | 12 | |||
BC054753.1 | - | 1 | 61 | 6411651 | ||
BC085270.1 | - | 1 | 54 | 6478784 | ||
BC157916.1 | - | 1 | 74 | 9007344 | ||
BU705914.1 | - | 1 | 245 | |||
BY275360.1 | - | 1 | 240 | |||
KJ650201.1 | - | 1 | 21 | |||
L26528.1 | - | 1 | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK133808.1 | - | 248 | Chr. - - [-] on Build |