Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181070.6 | NP_851415.1 | 2, 3 | 220 | ||
GenBank mRNA | AB232612.1 | - | 2, 3 | 122 | ||
AK045514.1 | - | 2, 3 | 183 | |||
AK140901.1 | - | 2, 3 | 188 | |||
AK143673.1 | - | 2, 3 | 196 | |||
AK146177.1 | - | 2, 3 | 145 | |||
AK146397.1 | - | 2, 3 | 145 | |||
AK154920.1 | - | 2, 3 | 197 | |||
BC056351.1 | - | 2, 3 | 174 | 6807284 | ||
L04966.1 | - | 2, 3 | 126 | |||
M79308.1 | - | 2, 3 | 122 | |||
X80333.1 | - | 2, 3 | 122 |