Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047789.1 | - | 16 | 1653 | ||
AK052567.1 | - | 17 | 1751 | |||
AK075948.1 | - | 17 | 1752 |
Gene Symbol: Kif20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166406.1 | NP_001159878.1 | 16 | 2311 | ||
NM_001166407.1 | NP_001159879.1 | 16 | 2309 | |||
NM_009004.4 | NP_033030.1 | 16 | 2205 | |||
GenBank mRNA | AK051201.1 | - | 16 | 2123 | ||
AK051995.1 | - | 16 | 2160 | |||
AK053280.1 | - | 16 | 2124 | |||
AK083412.1 | - | 16 | 2165 | |||
AK083697.1 | - | 16 | 2126 | |||
AK084732.1 | - | 16 | 2147 | |||
AK142764.1 | - | 16 | 2153 | |||
AK144844.1 | - | 16 | 2123 | |||
BC060608.1 | - | 16 | 2311 | 5720408 | ||
Y09632.1 | - | 16 | 2360 |