Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166406.1 | NP_001159878.1 | 9 | 1327 | ||
NM_001166407.1 | NP_001159879.1 | 9 | 1325 | |||
NM_009004.4 | NP_033030.1 | 9 | 1221 | |||
GenBank mRNA | AK051201.1 | - | 9 | 1139 | ||
AK051995.1 | - | 9 | 1176 | |||
AK053280.1 | - | 9 | 1140 | |||
AK083412.1 | - | 9 | 1181 | |||
AK083697.1 | - | 9 | 1140 | |||
AK084732.1 | - | 9 | 1163 | |||
AK142764.1 | - | 10 | 1071 | |||
AK144844.1 | - | 9 | 1139 | |||
BC060608.1 | - | 9 | 1327 | 5720408 | ||
Y09632.1 | - | 9 | 1376 |