Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290519.1 | NP_001277448.1 | 10 | 941 | ||
NM_001290520.1 | NP_001277449.1 | 8 | 864 | |||
NM_177576.3 | NP_808244.1 | 9 | 968 | |||
GenBank mRNA | AK052771.1 | - | 9 | 969 | ||
AK132922.1 | - | 10 | 942 | |||
HQ141857.1 | - | 10 | 931 |