Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009068.3 | NP_033094.3 | 2, 3 | 304 | ||
GenBank mRNA | AK030959.1 | - | 2, 3 | 535 | ||
AK048989.1 | - | 2, 3 | 532 | |||
AK156803.1 | - | 2, 3 | 569 | |||
BC050905.1 | - | 2, 3 | 304 | 5721177 | ||
BC054542.1 | - | 2, 3 | 304 | 5721177 | ||
BC058162.1 | - | 2, 3 | 490 | 5135235 | ||
U25995.1 | - | 1, 2 | 219 |