Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173483.1 | NP_001166954.1 | 7 | 1074 | ||
NM_020599.2 | NP_065624.1 | 7 | 1156 | |||
GenBank mRNA | AK014227.1 | - | 7 | 847 | ||
AK140622.1 | - | 7 | 1075 | |||
BC016268.1 | - | 6 | 749 | 4481973 |