Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011273.2 | NP_035403.1 | 13 | 1945 | ||
GenBank mRNA | AF114753.1 | - | 13 | 1830 | ||
AF131096.1 | - | 13 | 1735 | |||
AF198104.1 | - | 13 | 1735 | |||
AK048887.1 | - | 1 | 666 | |||
AK082228.1 | - | 1 | 1760 | |||
AK087140.1 | - | 4 | 1121 | |||
AK141660.1 | - | 13 | 1917 | |||
AK147403.1 | - | 13 | 1813 | |||
BC153872.1 | - | 13 | 1821 | 40110725 | ||
HQ323678.1 | - | 4 | 586 |
Set Membership: |
![]() |