Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011304.3 | NP_035434.1 | 14, 15 | 1566 | ||
GenBank mRNA | AB013912.1 | - | 14, 15 | 1397 | ||
AK135404.1 | - | 14, 15 | 1380 | |||
AK144156.1 | - | 14, 15 | 1388 | |||
AK159112.1 | - | 14, 15 | 1568 | |||
AK188084.1 | - | 1, 2 | 54 | |||
AK219298.1 | - | 1, 2 | 54 | |||
BC072585.1 | - | 2065 | ||||
BC115809.1 | - | 14, 15 | 1384 | 40105495 | ||
BC116693.1 | - | 14, 15 | 1383 | 40105493 |