Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011317.4 | NP_035447.3 | 6 | 1234 | ||
GenBank mRNA | AK044927.1 | - | 6 | 1231 | ||
AK050520.1 | - | 6 | 1229 | |||
AK146815.1 | - | 6 | 1196 | |||
AK150687.1 | - | 6 | 1213 | |||
AK152084.1 | - | 6 | 1213 | |||
AK204780.1 | - | 1 | 157 | |||
BC002051.1 | - | 6 | 1190 | 3490667 | ||
U17046.1 | - | 6 | 1191 | |||
U17961.1 | - | 6 | 1202 |