Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011317.4 | NP_035447.3 | 5 | 941 | ||
GenBank mRNA | AK037411.1 | - | 1 | 1975 | ||
AK044927.1 | - | 5 | 938 | |||
AK050520.1 | - | 5 | 936 | |||
AK146815.1 | - | 5 | 903 | |||
AK150687.1 | - | 5 | 920 | |||
AK152084.1 | - | 5 | 920 | |||
AK199670.1 | - | 3 | 264 | |||
BC002051.1 | - | 5 | 897 | 3490667 | ||
U17046.1 | - | 5 | 898 | |||
U17961.1 | - | 5 | 909 |