Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cx3cl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009142.3 | NP_033168.2 | 3 | 397 | ||
GenBank mRNA | AF010586.1 | - | 3 | 351 | ||
AF071549.1 | - | 3 | 388 | |||
AK040220.1 | - | 3 | 342 | |||
AK078285.1 | - | 3 | 375 | |||
AK138214.1 | - | 3 | 374 | |||
AK138472.1 | - | 3 | 374 | |||
AK138574.1 | - | 3 | 374 | |||
AK154646.1 | - | 3 | 374 | |||
BC006650.1 | - | 3 | 351 | 3498747 | ||
BC054838.1 | - | 3 | 357 | 6490082 | ||
CB526167.1 | - | 3 | 397 | |||
U92565.1 | - | 3 | 312 |