Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009148.3 | NP_033174.2 | 12, 13 | 1888 | ||
GenBank mRNA | AF022962.1 | - | 12, 13 | 1914 | ||
AK028250.1 | - | 10, 11 | 1620 | |||
AK173235.1 | - | 12, 13 | 1891 | |||
BC034644.1 | - | 12, 13 | 1888 | 5355276 |