Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009148.3 | NP_033174.2 | 5 | 732 | ||
GenBank mRNA | AF022962.1 | - | 5 | 758 | ||
AK010695.1 | - | 5 | 747 | |||
AK014499.1 | - | 5 | 755 | |||
AK028250.1 | - | 3 | 465 | |||
AK036507.1 | - | 5 | 744 | |||
AK038125.1 | - | 5 | 755 | |||
AK039983.1 | - | 5 | 755 | |||
AK086603.1 | - | 5 | 751 | |||
AK173235.1 | - | 5 | 735 | |||
AK188236.1 | - | 1 | 72 | |||
AK199139.1 | - | 2 | 111 | |||
AK212838.1 | - | 2 | 111 | |||
BC034644.1 | - | 5 | 732 | 5355276 |