Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sema6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130456.1 | NP_001123928.1 | 5, 6 | 535 | ||
NM_013662.2 | NP_038690.1 | 5, 6 | 646 | |||
GenBank mRNA | AF036585.1 | - | 4, 5 | 402 | ||
AK139358.1 | - | 5, 6 | 455 | |||
AK143229.1 | - | 5, 6 | 536 | |||
AK166077.1 | - | 5, 6 | 638 | |||
BC139415.1 | - | 4, 5 | 401 | 8862437 |