Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166450.2 | NP_001159922.1 | 4 | 591 | ||
NM_001290703.1 | NP_001277632.1 | 6 | 780 | |||
NM_001290704.1 | NP_001277633.1 | 6 | 771 | |||
NM_001290705.1 | NP_001277634.1 | 3 | 346 | |||
NM_011398.3 | NP_035528.1 | 4 | 678 | |||
GenBank mRNA | AF076981.1 | - | 4 | 679 | ||
AF155812.1 | - | 3 | 266 | |||
AF155813.1 | - | 3 | 257 | |||
AF535150.1 | - | 6 | 772 | |||
AF535151.1 | - | 6 | 781 | |||
AK148277.1 | - | 4 | 2126 | |||
BC048692.1 | - | 3 | 401 | 6333237 | ||
CX224035.1 | - | 5 | 529 | |||
CX238926.1 | - | 4 | 443 | |||
CX244278.1 | - | 4 | 677 |