Search Thermo Fisher Scientific
Gene Symbol: Slfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011408.1 | NP_035538.1 | 2 | 872 | ||
GenBank mRNA | AF099973.1 | - | 2 | 872 | ||
BC138749.1 | - | 2 | 827 | 8861771 | ||
BC138750.1 | - | 2 | 827 | 8861772 |