Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011420.2 | NP_035550.1 | 1, 2 | 116 | ||
GenBank mRNA | AK012698.1 | - | 1, 2 | 86 | ||
AK144778.1 | - | 1, 2 | 117 | |||
AK152495.1 | - | 1, 2 | 91 | |||
AK167832.1 | - | 1, 2 | 114 | |||
AY821787.1 | - | 1, 2 | 77 | |||
BC045158.1 | - | 1, 2 | 80 | 6486685 | ||
CJ058980.1 | - | 1, 2 | 117 | |||
U63294.1 | - | 1, 2 | 94 | |||
U77714.1 | - | 1, 2 | 77 | |||
Y12835.1 | - | 1, 2 | 102 |