Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013671.3 | NP_038699.2 | 5 | 831 | ||
GenBank mRNA | AK002428.1 | - | 5 | 691 | ||
AK002534.1 | - | 5 | 692 | |||
AK012354.1 | - | 5 | 689 | |||
AK135435.1 | - | 5 | 820 | |||
AK152047.1 | - | 5 | 689 | |||
AK167521.1 | - | 5 | 692 | |||
AK168082.1 | - | 5 | 692 | |||
AK168591.1 | - | 5 | 823 | |||
BC010548.1 | - | 5 | 667 | 3581909 | ||
BC018173.1 | - | 4 | 612 | |||
CT010273.1 | - | 5 | 624 | |||
X04972.1 | - | 5 | 676 | |||
Z18857.1 | - | 5 | 659 |