Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 16, 17 | 2449 | ||
GenBank mRNA | AB015790.1 | - | 16, 17 | 2273 | ||
AF031816.1 | - | 1727 | ||||
AK137635.1 | - | 4, 5 | 564 | |||
AK147303.1 | - | 16, 17 | 2450 | |||
AK155045.1 | - | 16, 17 | 2289 |