Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013672.2 | NP_038700.2 | 3 | 695 | ||
GenBank mRNA | AF022363.1 | - | 3 | 692 | ||
AF062566.1 | - | 3 | 631 |