Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177667.1 | NP_001171138.1 | 57 | 7617 | ||
NM_001177668.1 | NP_001171139.1 | 56 | 7557 | |||
GenBank mRNA | AK011566.1 | - | 15 | 1919 | ||
AK147317.1 | - | 56 | 7441 | |||
AK158599.1 | - | 19 | 2381 | |||
BC022120.1 | - | 26 | 3310 | |||
BC027791.1 | - | 33 | 4034 | |||
BC060504.1 | - | 12 | 1422 | |||
BC091776.1 | - | 12 | 1500 | |||
BC150941.1 | - | 57 | 7453 | 9087852 | ||
BC158015.1 | - | 57 | 7453 | 9007321 | ||
BC172095.1 | - | 56 | 7393 | 9054381 |