Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136081.2 | NP_001129553.1 | 2, 3 | 245 | ||
NM_182990.4 | NP_892035.2 | 2, 3 | 374 | |||
GenBank mRNA | AK146446.1 | - | 2, 3 | 297 | ||
AK146585.1 | - | 2, 3 | 294 | |||
AK146607.1 | - | 2, 3 | 277 | |||
AK146691.1 | - | 2, 3 | 277 | |||
AK147005.1 | - | 2, 3 | 307 | |||
AK151584.1 | - | 2, 3 | 327 | |||
AK158552.1 | - | 2, 3 | 245 | |||
BC004848.1 | - | 2, 3 | 241 | |||
BC042502.1 | - | 2, 3 | 222 | 5005209 | ||
CJ072796.1 | - | 376 | ||||
S50213.1 | - | 1, 2 | 119 |