Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077712.2 | NP_001071180.1 | 30 | 3807 | ||
NM_001290713.1 | NP_001277642.1 | 29 | 3533 | |||
NM_021465.4 | NP_067440.3 | 30 | 3667 | |||
GenBank mRNA | AJ002636.1 | - | 29 | 3509 | ||
AK034759.1 | - | 7 | 999 | |||
AK161072.1 | - | 2 | 273 | |||
BC066041.1 | - | 30 | 3548 | |||
CN708963.1 | - | 1 | 167 |