Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011506.3 | NP_035636.1 | 6 | 773 | ||
GenBank mRNA | AF058955.1 | - | 5 | 640 | ||
AK081965.1 | - | 6 | 794 | |||
AK088801.1 | - | 6 | 774 | |||
AK132762.1 | - | 6 | 763 | |||
AK150594.1 | - | 6 | 773 | |||
AK152783.1 | - | 6 | 773 | |||
AK153198.1 | - | 6 | 773 | |||
AK160130.1 | - | 6 | 774 | |||
AK160652.1 | - | 6 | 762 | |||
BC056353.1 | - | 6 | 764 | 6808360 | ||
BC057605.1 | - | 6 | 764 | 6808360 |