Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271724.1 | NP_001258653.2 | 3 | 387 | ||
NM_013677.2 | NP_038705.2 | 3 | 387 | |||
GenBank mRNA | AK002469.1 | - | 3 | 201 | ||
AK154726.1 | - | 3 | 207 | |||
BC004755.1 | - | 3 | 387 | 3584373 | ||
BC052500.1 | - | 3 | 374 | 5255619 | ||
BU697260.1 | - | 3 | 340 | |||
BY268659.1 | - | 3 | 243 |