Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145952.3 | NP_666064.3 | 5 | 1262 | ||
GenBank mRNA | AK172979.1 | - | 5 | 1256 | ||
BC138056.1 | - | 5 | 1158 | 8861075 | ||
BC138057.1 | - | 5 | 1158 | 8861076 |