Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011516.2 | NP_035646.2 | 6 | 425 | ||
GenBank mRNA | AK133229.1 | - | 6 | 426 | ||
BC137967.1 | - | 6 | 387 | 8860986 | ||
D88539.1 | - | 1 | 15 | |||
L41069.1 | - | 6 | 406 | |||
Z38118.1 | - | 6 | 416 |