Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011516.2 | NP_035646.2 | 26 | 2261 | ||
GenBank mRNA | AK076780.1 | - | 5 | 351 | ||
AK133229.1 | - | 26 | 2261 | |||
BC137967.1 | - | 26 | 2223 | 8860986 | ||
D88539.1 | - | 21 | 1851 | |||
L41069.1 | - | 26 | 2242 | |||
Z38118.1 | - | 26 | 2252 |