Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145952.1 | NP_001139424.1 | 6 | 724 | ||
NM_178665.5 | NP_848780.3 | 7 | 931 | |||
GenBank mRNA | AK029335.1 | - | 7 | 723 | ||
AK040185.1 | - | 2 | 247 | |||
AK040643.1 | - | 5 | 485 | |||
AK045288.1 | - | 7 | 792 | |||
AK045341.1 | - | 7 | 775 | |||
AK051345.1 | - | 7 | 803 | |||
AK051937.1 | - | 7 | 767 | |||
AK054550.1 | - | 6 | 727 | |||
AK085626.1 | - | 7 | 923 | |||
AK086307.1 | - | 7 | 742 | |||
AK168431.1 | - | 7 | 837 | |||
AK169780.1 | - | 7 | 743 | |||
BC085321.1 | - | 6 | 670 | 30743832 | ||
CG784727.1 | - | 3 | 315 |