Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145952.1 | NP_001139424.1 | 4 | 473 | ||
NM_178665.5 | NP_848780.3 | 5 | 680 | |||
GenBank mRNA | AK029335.1 | - | 5 | 472 | ||
AK040643.1 | - | 3 | 234 | |||
AK045288.1 | - | 5 | 541 | |||
AK045341.1 | - | 5 | 524 | |||
AK051345.1 | - | 5 | 552 | |||
AK051937.1 | - | 5 | 519 | |||
AK054550.1 | - | 4 | 476 | |||
AK085626.1 | - | 5 | 672 | |||
AK086307.1 | - | 5 | 491 | |||
AK168431.1 | - | 5 | 586 | |||
AK169780.1 | - | 5 | 492 | |||
BC085321.1 | - | 4 | 419 | 30743832 |